siRNA Details
| Gene Symbol: AU022754 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040433.1 | 2 | 580 | |||
| GenBank mRNA | AK159003.1 | - | 2 | 581 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB627747.1 | - | 3 | 499 | Chr. 15 - 85580252 - 85582707 [+] on Build GRCm38 | |

AU022754
Genomic Map