siRNA Details
| Gene Symbol: AU022754 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040433.1 | 2 | 916 | |||
| GenBank mRNA | AK159003.1 | - | 2 | 917 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AU022754| Gene Symbol: AU022754 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040433.1 | 2 | 916 | |||
| GenBank mRNA | AK159003.1 | - | 2 | 917 | ||