siRNA Details
| Gene Symbol: Gm4710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033456.1 | 1 | 88 | |||
| GenBank mRNA | AK082440.1 | - | 1 | 89 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB650123.1 | - | 1 | 89 | Chr. 17 - 75861023 - 75886387 [+] on Build GRCm38 | |

Gm4710
Genomic Map