siRNA Details
| Gene Symbol: Gm4710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033456.1 | 5 | 827 | |||
| GenBank mRNA | AK082440.1 | - | 5 | 828 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4710| Gene Symbol: Gm4710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033456.1 | 5 | 827 | |||
| GenBank mRNA | AK082440.1 | - | 5 | 828 | ||