siRNA Details
| Gene Symbol: A830008E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137140.1 | - | 1 | 895 | ||
| AK154552.1 | - | 4 | 3702 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A830008E24Rik| Gene Symbol: A830008E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137140.1 | - | 1 | 895 | ||
| AK154552.1 | - | 4 | 3702 | |||