siRNA Details
| Gene Symbol: Gm29685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK147773.1 | - | 305 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY740883.1 | - | 3, 4 | 302 | Chr. 10 - 107946647 - 107976215 [+] on Build GRCm38 | |

Gm29685
Genomic Map