siRNA Details
| Gene Symbol: 4833404L02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014654.1 | - | 1 | 538 | ||
| AK029357.1 | - | 1 | 537 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV248175.2 | - | 1 | 621 | Chr. 7 - 17058566 - 17059423 [+] on Build GRCm38 | |

4833404L02Rik
Genomic Map