siRNA Details
| Gene Symbol: C430002E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040385.1 | 2 | 961 | |||
| GenBank mRNA | AK082869.1 | - | 2 | 902 | ||
| AK145605.1 | - | 2 | 952 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C430002E04Rik| Gene Symbol: C430002E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040385.1 | 2 | 961 | |||
| GenBank mRNA | AK082869.1 | - | 2 | 902 | ||
| AK145605.1 | - | 2 | 952 | |||