siRNA Details
| Gene Symbol: Eny2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016562.1 | - | 1 | 461 | ||
| AK087816.1 | - | 1 | 461 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU518875.1 | - | 1 | 16 | Chr. 15 - 44428591 - 44429405 [+] on Build GRCm38 | |

Eny2
Genomic Map