siRNA Details
| Gene Symbol: Eny2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016562.1 | - | 1 | 654 | ||
| AK087816.1 | - | 1 | 655 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eny2| Gene Symbol: Eny2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016562.1 | - | 1 | 654 | ||
| AK087816.1 | - | 1 | 655 | |||