siRNA Details
| Gene Symbol: 4931430N09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046053.1 | 1 | 820 | |||
| GenBank mRNA | AK016492.1 | - | 1 | 822 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4931430N09Rik| Gene Symbol: 4931430N09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046053.1 | 1 | 820 | |||
| GenBank mRNA | AK016492.1 | - | 1 | 822 | ||