siRNA Details
| Gene Symbol: 0610031O16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC044770.1 | - | 348 | |||
| BC046436.1 | - | 348 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI144451.1 | - | 3 | 349 | Chr. 3 - 138229834 - 138240405 [-] on Build GRCm38 | |

0610031O16Rik
Genomic Map