siRNA Details
| Gene Symbol: Gm34433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC044776.2 | - | 4 | 1475 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm34433| Gene Symbol: Gm34433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC044776.2 | - | 4 | 1475 | ||