siRNA Details
| Gene Symbol: Gm34433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC044776.2 | - | 4 | 460 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI732276.1 | - | 4 | 461 | Chr. 7 - 19333129 - 19339181 [+] on Build GRCm38 | |

Gm34433
Genomic Map