siRNA Details
| Gene Symbol: 2810408I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038009.1 | 2 | 618 | |||
| GenBank mRNA | AK013046.1 | - | 2 | 619 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810408I11Rik| Gene Symbol: 2810408I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038009.1 | 2 | 618 | |||
| GenBank mRNA | AK013046.1 | - | 2 | 619 | ||