siRNA Details
| Gene Symbol: 2810408I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038009.1 | 2 | 675 | |||
| GenBank mRNA | AK013046.1 | - | 2 | 676 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA464200.1 | - | 2 | 615 | Chr. 1 - 64679864 - 64690596 [-] on Build GRCm38 | |

2810408I11Rik
Genomic Map