siRNA Details
| Gene Symbol: 4933431M02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017013.1 | - | 2 | 632 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716697.1 | - | 2 | 632 | Chr. 6 - 107627774 - 107632058 [+] on Build GRCm38 | |

4933431M02Rik
Genomic Map