siRNA Details
| Gene Symbol: 2810468N07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045176.1 | 2 | 1277 | |||
| GenBank mRNA | AK079380.1 | - | 2 | 1171 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810468N07Rik| Gene Symbol: 2810468N07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045176.1 | 2 | 1277 | |||
| GenBank mRNA | AK079380.1 | - | 2 | 1171 | ||