siRNA Details
| Gene Symbol: Asb17os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040373.1 | 2 | 200 | |||
| GenBank mRNA | AK010897.1 | - | 2 | 179 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asb17os| Gene Symbol: Asb17os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040373.1 | 2 | 200 | |||
| GenBank mRNA | AK010897.1 | - | 2 | 179 | ||