siRNA Details
| Gene Symbol: Gabra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036049.1 | - | 544 | |||
| AK039292.1 | - | 541 | ||||
| AK048165.1 | - | 3646 | ||||
| AK079593.1 | - | 750 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE650342.1 | - | 1 | 245 | Chr. 5 - 70959368 - 70959885 [-] on Build GRCm38 | |

Gabra2
Genomic Map