siRNA Details
| Gene Symbol: Gabra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036049.1 | - | 691 | |||
| AK048165.1 | - | 3793 | ||||
| AK079593.1 | - | 897 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabra2| Gene Symbol: Gabra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036049.1 | - | 691 | |||
| AK048165.1 | - | 3793 | ||||
| AK079593.1 | - | 897 | ||||