siRNA Details
| Gene Symbol: Qk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033279.1 | - | 1 | 1493 | ||
| AK168411.1 | - | 1 | 1648 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB478042.2 | - | 1 | 29 | Chr. 17 - 10202858 - 10203501 [-] on Build GRCm38 | |

Qk
Genomic Map