siRNA Details
| Gene Symbol: Gm36799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039939.1 | - | 1846 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm36799| Gene Symbol: Gm36799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039939.1 | - | 1846 | |||