siRNA Details
| Gene Symbol: Gm3510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138389.1 | - | 322 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY724079.1 | - | 2 | 318 | Chr. 5 - 15924555 - 15933403 [-] on Build GRCm38 | |

Gm3510
Genomic Map