siRNA Details
| Gene Symbol: Gm14169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040372.1 | 2 | 337 | |||
| GenBank mRNA | AK032272.1 | - | 2 | 338 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB622598.1 | - | 2 | 338 | Chr. 2 - 156611071 - 156613422 [-] on Build GRCm38 | |

Gm14169
Genomic Map