siRNA Details
| Gene Symbol: 4930528H21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019707.1 | - | 1 | 64 | ||
| BY715486.1 | - | 1 | 64 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930528H21Rik| Gene Symbol: 4930528H21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019707.1 | - | 1 | 64 | ||
| BY715486.1 | - | 1 | 64 | |||