siRNA Details
| Gene Symbol: Gm13049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033051.1 | - | 112 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB624029.1 | - | 1 | 112 | Chr. 4 - 150825733 - 150839484 [+] on Build GRCm38 | |

Gm13049
Genomic Map