siRNA Details
| Gene Symbol: Gm13049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033051.1 | - | 709 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV360674.2 | - | 3 | 636 | Chr. 4 - 150825808 - 150839577 [+] on Build GRCm38 | |

Gm13049
Genomic Map