siRNA Details
| Gene Symbol: 8430419K02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018427.1 | - | 4 | 1074 | ||
| BC050140.1 | - | 5 | 744 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF580972.1 | - | 5 | 784 | Chr. 11 - 18846582 - 18872161 [-] on Build GRCm38 | |

8430419K02Rik
Genomic Map