siRNA Details
| Gene Symbol: 8030448I15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020202.1 | - | 1 | 173 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720425.1 | - | 1 | 170 | Chr. 1 - 157156086 - 157156666 [-] on Build GRCm38 | |

8030448I15Rik
Genomic Map