siRNA Details
| Gene Symbol: 8030450C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020204.1 | - | 1 | 470 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720428.1 | - | 1 | 466 | Chr. 10 - 61931660 - 61932320 [-] on Build GRCm38 | |

8030450C14Rik
Genomic Map