siRNA Details
| Gene Symbol: 8030455M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020209.1 | - | 2, 3 | 390 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720434.1 | - | 2, 3 | 389 | Chr. 8 - 110047432 - 110056940 [+] on Build GRCm38 | |

8030455M16Rik
Genomic Map