siRNA Details
| Gene Symbol: Tmem86b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020752.1 | - | 385 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem86b| Gene Symbol: Tmem86b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020752.1 | - | 385 | |||