siRNA Details
| Gene Symbol: Gm20759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046030.1 | 5 | 833 | |||
| GenBank mRNA | AK135737.1 | - | 5 | 834 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20759| Gene Symbol: Gm20759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046030.1 | 5 | 833 | |||
| GenBank mRNA | AK135737.1 | - | 5 | 834 | ||