siRNA Details
| Gene Symbol: Gm20759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046030.1 | 5 | 542 | |||
| GenBank mRNA | AK135737.1 | - | 5 | 543 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ052541.1 | - | 5 | 543 | Chr. 11 - 86002914 - 86024350 [-] on Build GRCm38 | |

Gm20759
Genomic Map