siRNA Details
| Gene Symbol: 4933434C23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017052.1 | - | 1 | 209 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716734.1 | - | 1 | 209 | Chr. X - 117709406 - 117710207 [+] on Build GRCm38 | |

4933434C23Rik
Genomic Map