siRNA Details
| Gene Symbol: Mettl5os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050748.1 | - | 1 | 797 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB438887.2 | - | 1 | 443 | Chr. 2 - 69886012 - 69886640 [+] on Build GRCm38 | |

Mettl5os
Genomic Map