siRNA Details
| Gene Symbol: Gm11820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139819.1 | - | 3, 4 | 601 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY725908.1 | - | 3, 4 | 601 | Chr. 4 - 13076140 - 13096806 [+] on Build GRCm38 | |

Gm11820
Genomic Map