siRNA Details
| Gene Symbol: Gm9899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040427.1 | 3, 4 | 426 | |||
| GenBank mRNA | AK035982.1 | - | 3, 4 | 283 | ||
| AK138331.1 | - | 3, 4 | 429 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9899| Gene Symbol: Gm9899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040427.1 | 3, 4 | 426 | |||
| GenBank mRNA | AK035982.1 | - | 3, 4 | 283 | ||
| AK138331.1 | - | 3, 4 | 429 | |||