siRNA Details
| Gene Symbol: Gm9899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040427.1 | 4 | 611 | |||
| GenBank mRNA | AK035982.1 | - | 4 | 469 | ||
| AK138331.1 | - | 4 | 614 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY723983.1 | - | 4 | 614 | Chr. 5 - 30578134 - 30588619 [-] on Build GRCm38 | |

Gm9899
Genomic Map