siRNA Details
| Gene Symbol: Gm41409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK163292.1 | - | 176 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY725575.1 | - | 2 | 171 | Chr. 16 - 3764550 - 3785312 [+] on Build GRCm38 | |

Gm41409
Genomic Map