siRNA Details
Gene Symbol: Gm10612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158469.1 | - | 1, 2 | 155 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY753225.1 | - | 1, 2 | 155 | Chr. 8 - 122697860 - 122699152 [+] on Build GRCm38 |
