siRNA Details
| Gene Symbol: Gm10612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158469.1 | - | 2 | 233 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY753225.1 | - | 2 | 233 | Chr. 8 - 122697860 - 122699152 [+] on Build GRCm38 | |

Gm10612
Genomic Map