siRNA Details
| Gene Symbol: 2700088M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012587.1 | - | 1 | 70 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY711729.1 | - | 1 | 68 | Chr. 11 - 96294856 - 96295407 [+] on Build GRCm38 | |

2700088M07Rik
Genomic Map