siRNA Details
Gene Symbol: Gm15908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105041.1 | 3, 4 | 527 | |||
GenBank mRNA | AK132729.1 | - | 3, 4 | 528 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm15908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105041.1 | 3, 4 | 527 | |||
GenBank mRNA | AK132729.1 | - | 3, 4 | 528 |