siRNA Details
| Gene Symbol: Gm15908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105041.1 | 3 | 493 | |||
| GenBank mRNA | AK132729.1 | - | 3 | 494 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15908| Gene Symbol: Gm15908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105041.1 | 3 | 493 | |||
| GenBank mRNA | AK132729.1 | - | 3 | 494 | ||