siRNA Details
| Gene Symbol: 1700100I10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007101.1 | - | 2 | 742 | ||
| AK015607.1 | - | 5 | 965 | |||
| AV268330.1 | - | 1 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700100I10Rik| Gene Symbol: 1700100I10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007101.1 | - | 2 | 742 | ||
| AK015607.1 | - | 5 | 965 | |||
| AV268330.1 | - | 1 | 78 | |||