siRNA Details
| Gene Symbol: 4930480K23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015602.1 | - | 1 | 475 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715154.1 | - | 1 | 475 | Chr. 14 - 69732834 - 69733641 [+] on Build GRCm38 | |

4930480K23Rik
Genomic Map