siRNA Details
| Gene Symbol: 4930539N22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040598.1 | 4 | 672 | |||
| GenBank mRNA | AK016003.1 | - | 4 | 674 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715581.1 | - | 4 | 674 | Chr. 3 - 13597262 - 13652867 [-] on Build GRCm38 | |

4930539N22Rik
Genomic Map