siRNA Details
| Gene Symbol: Gm15894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038870.1 | - | 2018 | |||
| Gene Symbol: AW743432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AW822077.1 | - | 1 | 166 | ||

Gm15894
Genomic Map